Assay Details
| Gene Symbol: NDUFB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133578.4 | XP_003133626.1 | 155 | 65 | ||
| XM_005652448.3 | XP_005652505.1 | 155 | 65 | |||
| XM_021075850.1 | XP_020931509.1 | 155 | 65 | |||
| XM_021075852.1 | XP_020931511.1 | 155 | 65 | |||
| GenBank mRNA | AK230845.1 | - | 1 - 2 | 33 | 65 | |
| AK399697.1 | - | 1 - 2 | 53 | 65 | ||
| AY610245.1 | - | 1 - 2 | 30 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003133578.4, XM_005652448.3, XM_021075850.1, XM_021075852.1 transcripts.
Set Membership: |
|

NDUFB3
Genomic Map