Assay Details
| Gene Symbol: NDUFB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133578.4 | XP_003133626.1 | 238 | 100 | ||
| XM_005652449.3 | XP_005652506.1 | 312 | 100 | |||
| XM_021075847.1 | XP_020931506.1 | 298 | 100 | |||
| XM_021075848.1 | XP_020931507.1 | 226 | 100 | |||
| GenBank mRNA | AK230845.1 | - | 2 - 3 | 116 | 100 | |
| AK399697.1 | - | 2 - 3 | 136 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003133578.4, XM_005652449.3, XM_021075847.1, XM_021075848.1 transcripts.
Set Membership: |
|

NDUFB3
Genomic Map