Assay Details
| Gene Symbol: IRAK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091960.1 | XP_020947619.1 | 3021 | 100 | ||
| GenBank mRNA | AK230914.1 | - | 1 - 2 | 2128 | 100 | |
| AK345824.1 | - | 7 - 8 | 1081 | 100 | ||
| AK346429.1 | - | 7 - 8 | 811 | 100 | ||
| AK350251.1 | - | 7 - 8 | 866 | 100 | ||
More Information
Set Membership: |
|

IRAK3
Genomic Map