Assay Details
| Gene Symbol: SRP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121602.6 | XP_003121650.1 | 218 | 76 | ||
| GenBank mRNA | AK230923.1 | - | 2 - 3 | 151 | 76 | |
| AK392588.1 | - | 2 - 3 | 155 | 76 | ||
| AK399676.1 | - | 2 - 3 | 145 | 76 | ||
More Information
Set Membership: |
|

SRP14
Genomic Map