Assay Details
| Gene Symbol: EMR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083944.1 | XP_020939603.1 | 1776 | 78 | ||
| XM_021083945.1 | XP_020939604.1 | 1727 | 78 | |||
| XM_021083946.1 | XP_020939605.1 | 1777 | 78 | |||
| GenBank mRNA | AK230928.1 | - | 14 - 15 | 1622 | 78 | |
More Information
Set Membership: |
|

EMR4
Genomic Map