Assay Details
| Gene Symbol: GALK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925554.7 | XP_001925589.5 | 416 | 79 | ||
| XM_003121519.6 | XP_003121567.4 | 689 | 79 | |||
| XM_005659628.3 | XP_005659685.2 | 440 | 79 | |||
| XM_005659629.3 | XP_005659686.2 | 309 | 79 | |||
| XM_013993027.2 | XP_013848481.2 | 407 | 79 | |||
| XM_021073702.1 | XP_020929361.1 | 641 | 79 | |||
| XM_021073705.1 | XP_020929364.1 | 398 | 79 | |||
| XM_021073712.1 | XP_020929371.1 | 530 | 79 | |||
| GenBank mRNA | AK230940.1 | - | 3 - 4 | 342 | 79 | |
| AK343919.1 | - | 3 - 4 | 403 | 79 | ||
| AK346231.1 | - | 4 - 5 | 600 | 79 | ||
| AK393739.1 | - | 3 - 4 | 339 | 79 | ||
| AK399422.1 | - | 3 - 4 | 370 | 79 | ||
More Information
Set Membership: |
|

GALK2
Genomic Map