Assay Details
Gene Symbol: C14H12orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001929089.4 | XP_001929124.1 | 524 | 61 | ||
GenBank mRNA | AK235787.1 | - | 5 - 6 | 475 | 61 | |
AK238156.1 | - | 5 - 6 | 476 | 61 | ||
AK346318.1 | - | 5 - 6 | 470 | 61 | ||
AK398027.1 | - | 5 - 6 | 477 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
