Assay Details
| Gene Symbol: IARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086984.1 | XP_020942643.1 | 265 | 86 | ||
| XM_021086985.1 | XP_020942644.1 | 325 | 86 | |||
| XM_021086986.1 | XP_020942645.1 | 265 | 86 | |||
| XM_021086987.1 | XP_020942646.1 | 285 | 86 | |||
| GenBank mRNA | AK235793.1 | - | 2 - 3 | 292 | 86 | |
| AK352500.1 | - | 2 - 3 | 201 | 86 | ||
More Information
Set Membership: |
|

IARS1
Genomic Map