Assay Details
| Gene Symbol: IL17RD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021068996.1 | XP_020924655.1 | 853 | 73 | ||
| GenBank mRNA | AK235794.1 | - | 7 - 8 | 786 | 73 | |
More Information
Set Membership: |
|

IL17RD
Genomic Map