Assay Details
| Gene Symbol: COTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003361733.4 | XP_003361781.2 | 579 | 74 | ||
| GenBank mRNA | AK235815.1 | - | 3 - 4 | 507 | 74 | |
| AK239520.1 | - | 1 - 2 | 104 | 74 | ||
| AK393860.1 | - | 3 - 4 | 501 | 74 | ||
| AK394007.1 | - | 3 - 4 | 508 | 74 | ||
| AK395957.1 | - | 3 - 4 | 497 | 74 | ||
| AK397131.1 | - | 2 - 3 | 767 | 74 | ||
| AK400695.1 | - | 3 - 4 | 584 | 74 | ||
More Information
Set Membership: |
|

COTL1
Genomic Map