Assay Details
| Gene Symbol: PPIH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128098.4 | XP_003128146.2 | 151 | 72 | ||
| XM_003128099.4 | XP_003128147.2 | 151 | 72 | |||
| XM_003128100.4 | XP_003128148.2 | 151 | 72 | |||
| XM_021096918.1 | XP_020952577.1 | 75 | 72 | |||
| XM_021096919.1 | XP_020952578.1 | 75 | 72 | |||
| XM_021096920.1 | XP_020952579.1 | 151 | 72 | |||
| GenBank mRNA | AK235856.1 | - | 1 - 2 | 87 | 72 | |
| AK391754.1 | - | 1 - 2 | 66 | 72 | ||
| AY609841.1 | - | 1 - 2 | 73 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021096918.1, XM_021096919.1 transcripts.
Set Membership: |
|

PPIH
Genomic Map