Assay Details
Gene Symbol: GNL3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005673659.3 | XP_005673716.1 | 809 | 57 | ||
GenBank mRNA | AK235894.1 | - | 8 - 9 | 729 | 57 | |
AK389662.1 | - | 8 - 9 | 747 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
