Assay Details
| Gene Symbol: GPER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124244.5 | XP_003124292.1 | 166 | 98 | ||
| GenBank mRNA | AK235932.1 | - | 1 - 2 | 186 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003124244.5 transcripts.
Set Membership: |
|

GPER1
Genomic Map