Assay Details
| Gene Symbol: CSNK1G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021084209.1 | XP_020939868.1 | 1386 | 62 | ||
| XM_021084210.1 | XP_020939869.1 | 2064 | 62 | |||
| GenBank mRNA | AK235986.1 | - | 4 - 5 | 438 | 62 | |
More Information
Set Membership: |
|

CSNK1G2
Genomic Map