Assay Details
| Gene Symbol: WRAP53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131944.4 | XP_003131992.1 | 1576 | 57 | ||
| GenBank mRNA | AK236106.1 | - | 8 - 9 | 1514 | 57 | |
More Information
Set Membership: |
|

WRAP53
Genomic Map