Assay Details
| Gene Symbol: CREB3L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134635.4 | XP_003134683.1 | 6358 | 106 | ||
| GenBank mRNA | AK236199.1 | - | 1 | 416 | 106 | |
| AK348022.1 | - | 2 | 1212 | 106 | ||
| AK390214.1 | - | 1 | 1232 | 106 | ||
More Information
Set Membership: |
|

CREB3L2
Genomic Map