Assay Details
| Gene Symbol: ENPP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021074886.1 | XP_020930545.1 | 1097 | 65 | ||
| GenBank mRNA | AK236257.1 | - | 6 - 7 | 1102 | 65 | |
| AK345705.1 | - | 6 - 7 | 1053 | 65 | ||
| AK347685.1 | - | 6 - 7 | 1047 | 65 | ||
| AY609869.1 | - | 6 - 7 | 1032 | 65 | ||
More Information
Set Membership: |
|

ENPP6
Genomic Map