Assay Details
| Gene Symbol: TRMT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927396.5 | XP_001927431.2 | 717 | 87 | ||
| XM_005672681.3 | XP_005672738.2 | 735 | 87 | |||
| GenBank mRNA | AK236259.1 | - | 5 - 6 | 576 | 87 | |
| AK393024.1 | - | 2 - 3 | 137 | 87 | ||
| AK399195.1 | - | 5 - 6 | 705 | 87 | ||
More Information
Set Membership: |
|

TRMT6
Genomic Map