Assay Details
| Gene Symbol: GARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134806.6 | XP_003134854.3 | 1364 | 70 | ||
| GenBank mRNA | AK236282.1 | - | 9 - 10 | 1053 | 70 | |
| AK397381.1 | - | 9 - 10 | 1290 | 70 | ||
More Information
Set Membership: |
|

GARS1
Genomic Map