Assay Details
| Gene Symbol: TRAF3IP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924787.5 | XP_001924822.2 | 2071 | 101 | ||
| XM_003353287.4 | XP_003353335.1 | 4601 | 101 | |||
| XM_013992757.2 | XP_013848211.1 | 1700 | 101 | |||
| XM_021091285.1 | XP_020946944.1 | 2402 | 101 | |||
| XM_021091306.1 | XP_020946965.1 | 320 | 101 | |||
| GenBank mRNA | AK236283.1 | - | 7 - 8 | 1713 | 101 | |
| AK350270.1 | - | 7 - 8 | 1650 | 101 | ||
More Information
Set Membership: |
|

TRAF3IP2
Genomic Map