Assay Details
| Gene Symbol: RIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021099628.1 | XP_020955287.1 | 2529 | 82 | ||
| GenBank mRNA | AK236285.1 | - | 1 - 2 | 465 | 82 | |
| AK397160.1 | - | 1 - 2 | 517 | 82 | ||
More Information
Set Membership: |
|

RIN3
Genomic Map