Assay Details
Gene Symbol: RSBN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021102552.1 | XP_020958211.1 | 730 | 96 | ||
GenBank mRNA | AK236348.1 | - | 2 - 3 | 705 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
