Assay Details
| Gene Symbol: OGFOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928564.5 | XP_001928599.1 | 1114 | 82 | ||
| XM_021074073.1 | XP_020929732.1 | 591 | 82 | |||
| GenBank mRNA | AK236397.1 | - | 5 - 6 | 1059 | 82 | |
| AK351194.1 | - | 5 - 6 | 533 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021074073.1 transcripts.
Set Membership: |
|

OGFOD2
Genomic Map