Assay Details
| Gene Symbol: ANO10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013981530.2 | XP_013836984.2 | 1329 | 82 | ||
| XM_013981531.2 | XP_013836985.2 | 1254 | 82 | |||
| XM_021071717.1 | XP_020927376.1 | 1502 | 82 | |||
| XM_021071718.1 | XP_020927377.1 | 1431 | 82 | |||
| XM_021071719.1 | XP_020927378.1 | 1430 | 82 | |||
| XM_021071720.1 | XP_020927379.1 | 1329 | 82 | |||
| GenBank mRNA | AK236407.1 | - | 1 - 2 | 429 | 82 | |
More Information
Set Membership: |
|

ANO10
Genomic Map