Assay Details
| Gene Symbol: GALNT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021072437.1 | XP_020928096.1 | 251 | 60 | ||
| XM_021072438.1 | XP_020928097.1 | 251 | 60 | |||
| GenBank mRNA | AK236470.1 | - | 1 - 2 | 147 | 60 | |
| AK345056.1 | - | 1 - 2 | 176 | 60 | ||
More Information
Set Membership: |
|

GALNT7
Genomic Map