Assay Details
| Gene Symbol: ANGPT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013985210.2 | XP_013840664.1 | 3541 | 73 | ||
| GenBank mRNA | AK236555.1 | - | 2 - 3 | 607 | 73 | |
More Information
Set Membership: |
|

ANGPT4
Genomic Map