Assay Details
| Gene Symbol: PUS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928590.5 | XP_001928625.1 | 1257 | 88 | ||
| XM_003132854.5 | XP_003132902.2 | 1729 | 88 | |||
| XM_005670523.3 | XP_005670580.1 | 1344 | 88 | |||
| XM_005670524.3 | XP_005670581.1 | 1417 | 88 | |||
| XM_005670525.3 | XP_005670582.1 | 1337 | 88 | |||
| GenBank mRNA | AK236556.1 | - | 5 - 6 | 1202 | 88 | |
| AK236827.1 | - | 5 - 6 | 1215 | 88 | ||
| AK390841.1 | - | 5 - 6 | 1302 | 88 | ||
| AK396128.1 | - | 5 - 6 | 1312 | 88 | ||
More Information
Set Membership: |
|

PUS1
Genomic Map