Assay Details
| Gene Symbol: SSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924998.6 | XP_001925033.2 | 216 | 65 | ||
| XM_005671925.3 | XP_005671982.1 | 265 | 65 | |||
| GenBank mRNA | AK236611.1 | - | 2 - 3 | 168 | 65 | |
| AK393008.1 | - | 2 - 3 | 172 | 65 | ||
| AY609914.1 | - | 2 - 3 | 151 | 65 | ||
More Information
Set Membership: |
|

SSB
Genomic Map