Assay Details
| Gene Symbol: PAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070136.1 | XP_020925795.1 | 650 | 92 | ||
| XM_021070137.1 | XP_020925796.1 | 539 | 92 | |||
| XM_021070138.1 | XP_020925797.1 | 467 | 92 | |||
| GenBank mRNA | AK236653.1 | - | 3 - 4 | 559 | 92 | |
| AK348141.1 | - | 3 - 4 | 559 | 92 | ||
| AK351583.1 | - | 3 - 4 | 535 | 92 | ||
More Information
Set Membership: |
|

PAK2
Genomic Map