Assay Details
| Gene Symbol: LOC100523244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126024.6 | XP_003126072.1 | 1545 | 95 | ||
| XM_005655506.3 | XP_005655563.1 | 1607 | 95 | |||
| XM_005655509.3 | XP_005655566.1 | 1542 | 95 | |||
| XM_021091492.1 | XP_020947151.1 | 1376 | 95 | |||
| GenBank mRNA | AK236660.1 | - | 10 - 11 | 1443 | 94 | |
| AK391789.1 | - | 10 - 11 | 1522 | 95 | ||
More Information
Set Membership: |
|

LOC100523244
Genomic Map