Assay Details
| Gene Symbol: LOC100523244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005655509.3 | XP_005655566.1 | 258 | 57 | ||
| GenBank mRNA | AK236660.1 | - | 1 - 2 | 159 | 57 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005655509.3 transcripts.
Set Membership: |
|

LOC100523244
Genomic Map