Assay Details
| Gene Symbol: ZBTB8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356289.4 | XP_003356337.1 | 2063 | 85 | ||
| XM_005665187.3 | XP_005665244.1 | 1142 | 85 | |||
| GenBank mRNA | AK236688.1 | - | 4 - 5 | 1272 | 85 | |
More Information
Set Membership: |
|

ZBTB8A
Genomic Map