Assay Details
| Gene Symbol: CANT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066517.1 | XP_020922176.1 | 763 | 78 | ||
| GenBank mRNA | AK236692.1 | - | 2 - 3 | 700 | 78 | |
| AK351274.1 | - | 2 - 3 | 700 | 78 | ||
More Information
Set Membership: |
|

CANT1
Genomic Map