Assay Details
| Gene Symbol: PYCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090543.1 | XP_020946202.1 | 440 | 122 | ||
| GenBank mRNA | AK236710.1 | - | 3 - 4 | 365 | 122 | |
More Information
Set Membership: |
|

PYCR3
Genomic Map