Assay Details
| Gene Symbol: IMMP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021079086.1 | XP_020934745.1 | 294 | 72 | ||
| XM_021079087.1 | XP_020934746.1 | 403 | 72 | |||
| XM_021079088.1 | XP_020934747.1 | 294 | 72 | |||
| XM_021079089.1 | XP_020934748.1 | 294 | 72 | |||
| XM_021079090.1 | XP_020934749.1 | 294 | 72 | |||
| GenBank mRNA | AK236801.1 | - | 3 - 4 | 252 | 72 | |
| AK237369.1 | - | 2 - 3 | 445 | 72 | ||
More Information
Set Membership: |
|

IMMP2L
Genomic Map