Assay Details
| Gene Symbol: REEP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003480887.4 | XP_003480935.1 | 3929 | 108 | ||
| GenBank mRNA | AK236819.1 | - | 1 | 914 | 108 | |
| AK348980.1 | - | 1 | 986 | 108 | ||
| AK389736.1 | - | 1 | 1406 | 108 | ||
| AK398554.1 | - | 5 | 2981 | 108 | ||
| AK399492.1 | - | 1 | 831 | 108 | ||
More Information
Set Membership: |
|

REEP5
Genomic Map