Assay Details
| Gene Symbol: MOV10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005655442.3 | XP_005655499.1 | 2785 | 61 | ||
| XM_021090739.1 | XP_020946398.1 | 2428 | 61 | |||
| XM_021090741.1 | XP_020946400.1 | 2402 | 61 | |||
| GenBank mRNA | AK236859.1 | - | 4 - 5 | 537 | 61 | |
| AK239796.1 | - | 3 - 4 | 394 | 61 | ||
More Information
Set Membership: |
|

MOV10
Genomic Map