Assay Details
| Gene Symbol: TSPYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121374.4 | XP_003121422.2 | 848 | 88 | ||
| GenBank mRNA | AK236994.1 | - | 1 | 819 | 88 | |
| AK345257.1 | - | 1 | 864 | 88 | ||
| AK349198.1 | - | 1 | 845 | 88 | ||
More Information
Set Membership: |
|

TSPYL1
Genomic Map