Assay Details
| Gene Symbol: TENT4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094215.1 | XP_020949874.1 | 1243 | 132 | ||
| XM_021094216.1 | XP_020949875.1 | 1094 | 132 | |||
| XM_021094217.1 | XP_020949876.1 | 1250 | 132 | |||
| GenBank mRNA | AK237049.1 | - | 2 - 3 | 223 | 132 | |
More Information
Set Membership: |
|

TENT4B
Genomic Map