Assay Details
| Gene Symbol: MARCHF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091899.1 | XP_020947558.1 | 444 | 89 | ||
| GenBank mRNA | AK237073.1 | - | 1 - 2 | 184 | 89 | |
More Information
Set Membership: |
|

MARCHF9
Genomic Map