Assay Details
| Gene Symbol: FAM126B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK237084.1 | - | 2 - 3 | 104 | 91 | |
| AK238012.1 | - | 2 - 3 | 104 | 91 | ||
| AK344431.1 | - | 2 - 3 | 92 | 91 | ||
More Information
Set Membership: |
|

FAM126B
Genomic Map