Assay Details
| Gene Symbol: FBXO31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093683.1 | XP_020949342.1 | 3422 | 114 | ||
| XM_021093684.1 | XP_020949343.1 | 3341 | 114 | |||
| XM_021093687.1 | XP_020949346.1 | 3252 | 114 | |||
| XM_021093688.1 | XP_020949347.1 | 3061 | 114 | |||
| XM_021093689.1 | XP_020949348.1 | 3061 | 114 | |||
| XM_021093690.1 | XP_020949349.1 | 3060 | 114 | |||
| XM_021093691.1 | XP_020949350.1 | 3061 | 114 | |||
| XM_021093692.1 | XP_020949351.1 | 4956 | 114 | |||
| XM_021093693.1 | XP_020949352.1 | 3165 | 114 | |||
| GenBank mRNA | AK237185.1 | - | 1 | 1720 | 114 | |
More Information
Set Membership: |
|

FBXO31
Genomic Map