Assay Details
| Gene Symbol: PTP4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088812.1 | XP_020944471.1 | 1185 | 53 | ||
| XM_021088813.1 | XP_020944472.1 | 1513 | 53 | |||
| XM_021088814.1 | XP_020944473.1 | 1156 | 53 | |||
| XM_021088815.1 | XP_020944474.1 | 1027 | 53 | |||
| XM_021088816.1 | XP_020944475.1 | 1271 | 53 | |||
| XM_021088817.1 | XP_020944476.1 | 1163 | 53 | |||
| XM_021088818.1 | XP_020944477.1 | 1094 | 53 | |||
| GenBank mRNA | AK237414.1 | - | 2 - 3 | 292 | 53 | |
More Information
Set Membership: |
|

PTP4A3
Genomic Map