Assay Details
| Gene Symbol: PAX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122019.6 | XP_003122067.3 | 826 | 79 | ||
| XM_021066090.1 | XP_020921749.1 | 1045 | 79 | |||
| XM_021066095.1 | XP_020921754.1 | 1515 | 79 | |||
| XM_021066100.1 | XP_020921759.1 | 1193 | 79 | |||
| XM_021066104.1 | XP_020921763.1 | 1379 | 79 | |||
| GenBank mRNA | AK237481.1 | - | 3 - 4 | 888 | 79 | |
| AK345564.1 | - | 3 - 4 | 1020 | 79 | ||
| AK348896.1 | - | 456 | 78 | |||
| AK390845.1 | - | 3 - 4 | 1012 | 79 | ||
More Information
Set Membership: |
|

PAX5
Genomic Map