Assay Details
| Gene Symbol: PPP4R3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005666398.3 | XP_005666455.1 | 997 | 110 | ||
| XM_005666399.3 | XP_005666456.1 | 1031 | 110 | |||
| XM_021099618.1 | XP_020955277.1 | 311 | 110 | |||
| XM_021099619.1 | XP_020955278.1 | 311 | 110 | |||
| GenBank mRNA | AK237518.1 | - | 1 - 2 | 972 | 110 | |
| AK349358.1 | - | 1 - 2 | 972 | 110 | ||
| AK395808.1 | - | 1 - 2 | 990 | 110 | ||
More Information
Set Membership: |
|

PPP4R3A
Genomic Map