Assay Details
| Gene Symbol: CCDC189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013992406.2 | XP_013847860.2 | 511 | 68 | ||
| GenBank mRNA | AK237519.1 | - | 1 - 2 | 272 | 68 | |
More Information
Set Membership: |
|

CCDC189
Genomic Map