Assay Details
| Gene Symbol: DPY19L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021079309.1 | XP_020934968.1 | 2193 | 117 | ||
| XM_021079310.1 | XP_020934969.1 | 2091 | 117 | |||
| XM_021079311.1 | XP_020934970.1 | 2091 | 117 | |||
| XM_021079312.1 | XP_020934971.1 | 2091 | 117 | |||
| XM_021079313.1 | XP_020934972.1 | 2091 | 117 | |||
| XM_021079314.1 | XP_020934973.1 | 2091 | 117 | |||
| XM_021079315.1 | XP_020934974.1 | 2091 | 117 | |||
| XM_021079316.1 | XP_020934975.1 | 2091 | 117 | |||
| XM_021079317.1 | XP_020934976.1 | 2091 | 117 | |||
| XM_021079318.1 | XP_020934977.1 | 2091 | 117 | |||
| XM_021079319.1 | XP_020934978.1 | 2091 | 117 | |||
| XM_021079320.1 | XP_020934979.1 | 2091 | 117 | |||
| XM_021079321.1 | XP_020934980.1 | 2091 | 117 | |||
| XM_021079323.1 | XP_020934982.1 | 2091 | 117 | |||
| XM_021079324.1 | XP_020934983.1 | 2091 | 117 | |||
| XM_021079325.1 | XP_020934984.1 | 2188 | 117 | |||
| GenBank mRNA | AK237525.1 | - | 1 - 2 | 42 | 117 | |
More Information
Set Membership: |
|

DPY19L1
Genomic Map