Assay Details
| Gene Symbol: SRPRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021069470.1 | XP_020925129.1 | 302 | 64 | ||
| GenBank mRNA | AK237582.1 | - | 2 - 3 | 273 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SRPRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021069470.1 | XP_020925129.1 | 302 | 64 | ||
| GenBank mRNA | AK237582.1 | - | 2 - 3 | 273 | 64 | |
Set Membership: |
|