Assay Details
| Gene Symbol: THG1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134104.4 | XP_003134152.3 | 411 | 91 | ||
| GenBank mRNA | AK237675.1 | - | 2 - 3 | 389 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: THG1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134104.4 | XP_003134152.3 | 411 | 91 | ||
| GenBank mRNA | AK237675.1 | - | 2 - 3 | 389 | 91 | |
Set Membership: |
|