Assay Details
| Gene Symbol: ARHGAP45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003353961.4 | XP_003354009.1 | 2809 | 66 | ||
| XM_021084281.1 | XP_020939940.1 | 2858 | 66 | |||
| XM_021084282.1 | XP_020939941.1 | 2843 | 66 | |||
| XM_021084283.1 | XP_020939942.1 | 2544 | 66 | |||
| XM_021084284.1 | XP_020939943.1 | 2250 | 66 | |||
| GenBank mRNA | AK237727.1 | - | 3 - 4 | 406 | 66 | |
More Information
Set Membership: |
|

ARHGAP45
Genomic Map